Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PIP5K59B
Entrez Gene ID
37633
Gene Name
Phosphatidylinositol 4-phosphate 5-kinase 59B
Gene Aliases
CG17281, CG3682, Dmel\CG3682, Dmel_CG3682, PI4P5K, PIP5K 59B, dPIP5K, dPIP5K59B, dpip5k
Chromosome Location
Chr.2R: 22874769 - 22881436 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0034789
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001043101.4 NP_001036566.4 2 - 3 387 74
NM_001259542.2 NP_001246471.1 2 - 3 387 74
NM_001259543.2 NP_001246472.1 2 - 3 387 74
NM_001382080.1 NP_001369113.1 2 - 3 387 74
NM_137885.4 NP_611729.2 2 - 3 387 74
NM_166567.4 NP_726262.2 2 - 3 387 74
GenBank mRNA BT044218.1 - 2 - 3 366 74

View Full Product Details