Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
yip3
Entrez Gene ID
37683
Gene Name
yippee interacting protein 3
Gene Aliases
BcDNA:AT20887, CG13549, Dmel\CG13549, Dmel_CG13549, NEST:bs18b12, bs18b12.y1
Chromosome Location
Chr.2R: 23244044 - 23245399 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0040063
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_079095.4 NP_523819.2 3 - 4 234 107
NM_166588.3 NP_726300.1 2 - 3 695 107
GenBank mRNA BT044391.1 - 2 - 3 585 107

View Full Product Details