Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ord
Entrez Gene ID
37687
Gene Name
orientation disruptor
Gene Aliases
CG3134, Dmel\CG3134, Dmel_CG3134, ORD, Ord
Chromosome Location
Chr.2R: 23271257 - 23275536 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0003009
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001299831.1 NP_001286760.1 1 - 2 82 112
NM_166589.2 NP_726303.1 1 - 2 1819 112
GenBank mRNA BT122096.2 - 1 - 2 57 112

View Full Product Details