Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Try29F
Entrez Gene ID
34226
Gene Name
Trypsin 29F
Gene Aliases
CG9564, Dmel\CG9564, Dmel_CG9564, SP106, try29F, try29F.2, trypsin29F
Chromosome Location
Chr.2L: 8998537 - 8999816 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0015316
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001298843.1 NP_001285772.1 1 - 1 848 89
NM_078794.3 NP_523518.2 1 - 1 848 89

View Full Product Details