Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Toll-4
- Entrez Gene ID
-
34235
- Gene Name
- CG18241-PA
- Gene Aliases
-
CG18241, CT29238, Dmel\CG18241, Dmel_CG18241, TL4, Tl-4, Toll, Toll4, dTLR4, dToll4, toll
- Chromosome Location
-
Chr.2L: 9084107 - 9089440 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0032095
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_078795.2 |
NP_523519.2 |
3 - 4 |
912 |
108 |
View Full Product Details