Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Or30a
- Entrez Gene ID
-
34236
- Gene Name
- Odorant receptor 30a
- Gene Aliases
-
30A.1, 30a, CG13106, DOR56, Dmel\CG13106, Dmel_CG13106
- Chromosome Location
-
Chr.2L: 9111814 - 9113285 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0032096
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_078796.3 |
NP_523520.2 |
2 - 3 |
454 |
113 |
View Full Product Details