Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nos
Entrez Gene ID
34495
Gene Name
Nitric oxide synthase
Gene Aliases
CG6713, DNOS, DNOS1, Dmel\CG6713, Dmel_CG6713, NOS, dNOS, dNos, drNOSoxy, iNOS, nNOS, nos
Chromosome Location
Chr.2L: 10804274 - 10837511 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0011676
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001032069.2 NP_001027240.1 12 - 13 2499 82
NM_001032072.2 NP_001027243.2 10 - 11 1803 82
NM_078817.4 NP_523541.2 12 - 13 2499 82
GenBank mRNA BT010016.1 - 12 - 13 2498 82

View Full Product Details