Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lar
Entrez Gene ID
35259
Gene Name
Leukocyte-antigen-related-like
Gene Aliases
CG10443, DLAR, DLar, Dlar, Dmel\CG10443, Dmel_CG10443, LAR, PTP38A, Ptp38A, dlar, lar
Chromosome Location
Chr.2L: 19586623 - 19732069 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0000464
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001259165.1 NP_001246094.1 9 - 10 1510 64
NM_001273665.1 NP_001260594.1 11 - 12 2288 64
NM_001273666.1 NP_001260595.1 10 - 11 2792 64
NM_001273667.1 NP_001260596.1 10 - 11 1937 64
NM_001273668.1 NP_001260597.1 11 - 12 2141 64
NM_078880.3 NP_523604.2 8 - 9 1464 64
NM_165313.4 NP_724220.2 9 - 10 1806 64

Gene Symbol
CG46244
Entrez Gene ID
26067072
Gene Name
CG46244-PA
Gene Aliases
Dm_2L:103252, Dmel\CG46244, Dmel_CG46244
Chromosome Location
Chr.2L: 19606569 - 19731561 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0267964
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001316402.1 NP_001303331.1 10 - 11 2792 64

View Full Product Details