Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Cyp6a2
- Entrez Gene ID
-
35587
- Gene Name
- Cytochrome P450-6a2
- Gene Aliases
-
143127_at, 6a2, 91-R, 91R, CG9438, CYP6A2, CYP6a2, Cyp6A2, Cyp6a, Cyt-P450-B1, Dmel\CG9438, Dmel_CG9438, P-450, P-450-B, P450, P450-B, P450-B1, cyp6a2
- Chromosome Location
-
Chr.2R: 6779749 - 6781491 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0000473
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_078904.2 |
NP_523628.1 |
1 - 2 |
1157 |
90 |
GenBank mRNA |
BT015971.1 |
- |
1 - 2 |
1157 |
90 |
View Full Product Details