Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tsp42Ec
Entrez Gene ID
35612
Gene Name
Tetraspanin 42Ec
Gene Aliases
CG12847, Dm.Tsp42Ec, Dmel\CG12847, Dmel_CG12847
Chromosome Location
Chr.2R: 7006442 - 7008622 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0033124
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078905.3 NP_523629.1 2 - 3 384 86
GenBank mRNA AF274016.1 - 2 - 3 379 86
AY118995.1 - 2 - 3 379 86

View Full Product Details