Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tsp42Ee
Entrez Gene ID
35614
Gene Name
Tetraspanin 42Ee
Gene Aliases
BcDNA:LD16071, CG10106, Dm.Tsp42Ee, Dmel\CG10106, Dmel_CG10106, anon-WO0118547.151
Chromosome Location
Chr.2R: 7012254 - 7018362 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0029506
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001273824.1 NP_001260753.1 3 - 4 516 110
NM_078907.3 NP_523631.1 3 - 4 529 110
GenBank mRNA AF220043.1 - 3 - 4 497 110
BT044107.1 - 3 - 4 527 110

View Full Product Details