Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tsp42Eq
Entrez Gene ID
35628
Gene Name
Tetraspanin 42Eq
Gene Aliases
CG12832, Dm.Tsp42Eq, Dmel\CG12832, Dmel_CG12832
Chromosome Location
Chr.2R: 7062108 - 7063569 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0033138
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078919.3 NP_523643.2 2 - 3 309 58
GenBank mRNA BT150209.1 - 2 - 3 306 58

View Full Product Details