Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ptc
- Entrez Gene ID
-
35851
- Gene Name
- patched
- Gene Aliases
-
BcDNA:RH36596, CG2411, Conf, Dmel\CG2411, Dmel_CG2411, PTC, Ptc, Ptc_Dm, Ptch, com4, dPtc, l(2)k02507, pat, ptch, rubr, tuf
- Chromosome Location
-
Chr.2R: 8649649 - 8665223 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0003892
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_078937.4 |
NP_523661.2 |
3 - 4 |
3385 |
61 |
View Full Product Details