Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Or45b
- Entrez Gene ID
-
35980
- Gene Name
- Odorant receptor 45b
- Gene Aliases
-
45F.1, 45b, CG12931, DOR107, Dmel\CG12931, Dmel_CG12931
- Chromosome Location
-
Chr.2R: 9559598 - 9561068 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0033422
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_078943.1 |
NP_523667.1 |
2 - 3 |
715 |
61 |
View Full Product Details