Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Def
Entrez Gene ID
36047
Gene Name
Defensin
Gene Aliases
143607_at, CG1385, DEF, DIM 26, Dfn, Dmel\CG1385, Dmel_CG1385, def, defensin
Chromosome Location
Chr.2R: 10054178 - 10054576 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0010385
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078948.3 NP_523672.1 1 - 1 78 101
GenBank mRNA BT023384.1 - 1 - 1 78 101
KX531302.1 - 1 - 1 69 101
Z27247.1 - - 2771 101

View Full Product Details