Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rho-7
Entrez Gene ID
36281
Gene Name
rhomboid-7
Gene Aliases
CG8972, Dmel\CG8972, Dmel_CG8972, PARL, R7, Rho-7, Rhomboid-7, anon-WO0138581.3
Chromosome Location
Chr.2R: 11986513 - 11988787 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0033672
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001299395.1 NP_001286324.1 1 - 2 167 76
NM_078980.4 NP_523704.1 1 - 2 167 76

View Full Product Details