Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Gbeta76C
- Entrez Gene ID
-
40148
- Gene Name
- G protein beta-subunit 76C
- Gene Aliases
-
CG8770, DGqbeta, Dmel\CG8770, Dmel_CG8770, G&bgr76C, G[[beta76C]], G[[beta]], G[[beta]]e, G[q], Galpha76C, Gbe, Gbeta, Gbeta76c, Gbeta[[e]], Gbetae, Gbeta{e}, Gqbeta, Gqbetae, eGbeta, eye Gbeta, gbe
- Chromosome Location
-
Chr.3L: 19687637 - 19689992 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0004623
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_078996.3 |
NP_523720.2 |
1 - 2 |
158 |
75 |
GenBank mRNA |
AY118886.1 |
- |
1 - 2 |
129 |
75 |
View Full Product Details