Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- vg
- Entrez Gene ID
-
36421
- Gene Name
- vestigial
- Gene Aliases
-
CG3830, Dmel\CG3830, Dmel_CG3830, VG, Vg, vg21
- Chromosome Location
-
Chr.2R: 12884632 - 12899385 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0003975
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_078999.4 |
NP_523723.1 |
2 - 3 |
1552 |
68 |
View Full Product Details