Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- RagC-D
- Entrez Gene ID
-
35793
- Gene Name
- Ras-related GTP binding C/D
- Gene Aliases
-
CG8707, Dmel\CG8707, Dmel_CG8707, Gtr2, RagC, dRagC, ragC
- Chromosome Location
-
Chr.2R: 8161679 - 8163500 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0033272
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_136517.3 |
NP_610361.1 |
3 - 4 |
867 |
69 |
GenBank mRNA |
AY051510.1 |
- |
3 - 4 |
842 |
69 |
View Full Product Details