Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cyp6a13
Entrez Gene ID
35837
Gene Name
CG2397-PA
Gene Aliases
6a13, CG2397, CYP6A13, Cyp6A13, Dmel\CG2397, Dmel_CG2397, cyp6a13
Chromosome Location
Chr.2R: 8570499 - 8572147 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0033304
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_136546.3 NP_610390.1 1 - 1 1558 119
GenBank mRNA AY051723.1 - 1 - 1 1533 119

View Full Product Details