Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- beta3GalTII
- Entrez Gene ID
-
35848
- Gene Name
- beta-1,3-Galactosyltransferase II
- Gene Aliases
-
CG8734, Dmel\CG8734, Dmel_CG8734, GalTII, dGAGbeta3GalTII, dbeta3GalTII
- Chromosome Location
-
Chr.2R: 8626181 - 8627611 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0033315
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_136555.3 |
NP_610399.1 |
1 - 1 |
1143 |
84 |
GenBank mRNA |
AB300567.1 |
- |
1 - 1 |
1116 |
84 |
View Full Product Details