Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tom7
Entrez Gene ID
35899
Gene Name
Translocase of outer membrane 7
Gene Aliases
BcDNA:AT23266, CG8226, Dmel\CG8226, Dmel_CG8226, dtom7
Chromosome Location
Chr.2R: 8951595 - 8952242 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0033357
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001299291.1 NP_001286220.1 1 - 2 301 137
NM_136589.3 NP_610433.1 1 - 2 179 137
GenBank mRNA AY070812.1 - 1 - 2 208 137

View Full Product Details