Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- shrb
- Entrez Gene ID
-
35933
- Gene Name
- shrub
- Gene Aliases
-
CG8055, CHMP4C, DmSNF2, Dmel\CG8055, Dmel_CG8055, Dvps32, ESCRT, ESCRT-III, Shrb, Shrub, Snf7, Snf7/Vps32, Vps32, anon-WO0118547.180, vps32, vps32/snf7
- Chromosome Location
-
Chr.2R: 9142146 - 9143914 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0086656
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_136618.5 |
NP_610462.3 |
2 - 3 |
476 |
63 |
GenBank mRNA |
AY069131.1 |
- |
2 - 3 |
459 |
63 |
View Full Product Details