Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cyp4p3
Entrez Gene ID
35948
Gene Name
CG10843-PA
Gene Aliases
4p3, CG10843, Dmel\CG10843, Dmel_CG10843
Chromosome Location
Chr.2R: 9242320 - 9244410 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0033397
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_136629.3 NP_610473.1 3 - 4 523 101
GenBank mRNA AY075201.1 - 3 - 4 502 101

View Full Product Details