Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Camta
Entrez Gene ID
35974
Gene Name
Calmodulin-binding transcription activator
Gene Aliases
CG13952, CG13953, CG42332, CG8809, DmCAMTA, Dmel\CG42332, Dmel_CG42332, camta, cro, dCAMTA
Chromosome Location
Chr.2R: 9446046 - 9480076 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0287478
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144154.2 NP_001137626.1 1 - 1 284 58
NM_136647.5 NP_610491.4 1 - 1 284 58

View Full Product Details