Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Git
- Entrez Gene ID
-
36122
- Gene Name
- G protein-coupled receptor kinase interacting ArfGAP
- Gene Aliases
-
CG16728, CT37209, Dmel\CG16728, Dmel_CG16728, GIT, arfgap2, dGit, dgit, git
- Chromosome Location
-
Chr.2R: 10641818 - 10645427 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0033539
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_136755.4 |
NP_610599.3 |
3 - 4 |
1144 |
100 |
GenBank mRNA |
AY061380.1 |
- |
3 - 4 |
1133 |
100 |
View Full Product Details