Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- mms4
- Entrez Gene ID
-
36136
- Gene Name
- Methyl methanesulfonate sensitivity 4
- Gene Aliases
-
CG12936, Dmel\CG12936, Dmel_CG12936, EME1
- Chromosome Location
-
Chr.2R: 10811078 - 10812215 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0033549
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_136767.3 |
NP_610611.1 |
1 - 1 |
856 |
89 |
View Full Product Details