Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sha
Entrez Gene ID
36213
Gene Name
shavenoid
Gene Aliases
CG13209, Dmel\CG13209, Dmel_CG13209, Sha, koj, lincRNA.286
Chromosome Location
Chr.2R: 11324314 - 11336463 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0003382
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144173.3 NP_001137645.1 10 - 11 2443 70
NM_001273960.1 NP_001260889.1 8 - 9 2201 70
NM_136828.3 NP_610672.1 8 - 9 2225 70

View Full Product Details