Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lambdaTry
Entrez Gene ID
36214
Gene Name
CG12350-PA
Gene Aliases
CG12350, Dmel\CG12350, Dmel_CG12350, LambdaTry, SP76
Chromosome Location
Chr.2R: 11336840 - 11337791 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0043470
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_136829.4 NP_610673.1 1 - 1 889 136
GenBank mRNA AY070589.1 - 1 - 1 881 136

View Full Product Details