Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sobp
Entrez Gene ID
36260
Gene Name
Sine oculis-binding protein
Gene Aliases
CG8991, Dmel\CG8991, Dmel_CG8991, Sbp
Chromosome Location
Chr.2R: 11827170 - 11839825 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0033654
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001299391.1 NP_001286320.1 1 - 2 271 77
NM_136859.3 NP_610703.3 1 - 2 552 77
GenBank mRNA AY058444.1 - 1 - 2 257 77

View Full Product Details