Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- cuff
- Entrez Gene ID
-
36269
- Gene Name
- cutoff
- Gene Aliases
-
CG13190, Cuff, Dmel\CG13190, Dmel_CG13190
- Chromosome Location
-
Chr.2R: 11886067 - 11887603 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0260932
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_136865.3 |
NP_610709.1 |
1 - 2 |
575 |
82 |
View Full Product Details