Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cyp6g2
Entrez Gene ID
36317
Gene Name
CG8859-PA
Gene Aliases
6g2, CG8859, Cyp6G2, Dmel\CG8859, Dmel_CG8859
Chromosome Location
Chr.2R: 12188338 - 12190377 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0033696
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_136900.2 NP_610744.1 1 - 2 1287 72
GenBank mRNA BT031097.1 - 1 - 2 1287 72

View Full Product Details