Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dgt5
Entrez Gene ID
36364
Gene Name
dim gamma-tubulin 5
Gene Aliases
CG8828, Dgt5, Dmel\CG8828, Dmel_CG8828, msd2
Chromosome Location
Chr.2R: 12465678 - 12471775 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0033740
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001299420.1 NP_001286349.1 3 - 4 1678 101
NM_136941.3 NP_610785.1 4 - 5 1765 101
GenBank mRNA AY052018.1 - 3 - 4 1654 101

View Full Product Details