Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Phm
Entrez Gene ID
37823
Gene Name
Peptidylglycine-alpha-hydroxylating monooxygenase
Gene Aliases
CG3832, Dmel\CG3832, Dmel_CG3832, PHM, dPHM, lance-b, ll
Chromosome Location
Chr.2R: 23985175 - 23988455 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0283509
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_057877.4 NP_477225.1 5 - 6 699 81
NM_166644.2 NP_726394.1 5 - 6 699 81
GenBank mRNA AF006663.1 - 5 - 6 647 81
AY069103.1 - 5 - 6 665 81

View Full Product Details