Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tamo
Entrez Gene ID
37864
Gene Name
CG4057-PA
Gene Aliases
60B, CG4057, Dmel\CG4057, Dmel_CG4057
Chromosome Location
Chr.2R: 24126638 - 24133551 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0041582
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001299898.1 NP_001286827.1 2 - 3 721 72
NM_138045.2 NP_611889.1 2 - 3 411 72
NM_166664.1 NP_726426.1 2 - 3 721 72
GenBank mRNA BT001486.1 - 2 - 3 704 72

View Full Product Details