Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
prom
Entrez Gene ID
246493
Gene Name
prominin
Gene Aliases
CG30164, CG30165, CG30166, CG42310, CG4556, Dmel\CG42310, Dmel_CG42310, Prom, eyc
Chromosome Location
Chr.2R: 24415410 - 24428387 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0259210
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144281.2 NP_001137753.1 2 - 2 750 61
NM_001144282.2 NP_001137754.1 2 - 2 750 61
NM_001299914.1 NP_001286843.1 2 - 2 750 61
NM_166671.2 NP_726444.2 2 - 2 750 61
GenBank mRNA BT029279.1 - 2 - 2 652 61

View Full Product Details