Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pio
Entrez Gene ID
37929
Gene Name
piopio
Gene Aliases
CG30167, CG3541, Dmel\CG3541, Dmel_CG3541, Pio
Chromosome Location
Chr.2R: 24581245 - 24597862 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0020521
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169824.2 NP_001163295.1 2 - 3 894 95
NM_001299920.1 NP_001286849.1 2 - 3 140 95
NM_138094.4 NP_611938.2 3 - 4 418 95
NM_166677.3 NP_726461.1 3 - 4 422 95
GenBank mRNA AY862157.1 - 3 - 4 392 95
BT003795.1 - 2 - 3 877 95
BT011382.1 - 3 - 4 390 95

View Full Product Details