Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SiaT
Entrez Gene ID
37950
Gene Name
Sialyltransferase
Gene Aliases
CG4871, D.SiaT, D.SialT, DSiaT, Dm ST, DmPST, DmSialT, Dmel\CG4871, Dmel_CG4871, ST, ST6Gal, SialT, St6Gal, dST6Gal I, dSiaT
Chromosome Location
Chr.2R: 24691422 - 24694386 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0035050
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_079129.3 NP_523853.1 2 - 3 1466 87
NM_166684.2 NP_726474.1 3 - 4 846 87
GenBank mRNA AF218237.1 - 1 - 2 848 87
AF397532.1 - 2 - 3 809 87

View Full Product Details