Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SiaT
Entrez Gene ID
37950
Gene Name
Sialyltransferase
Gene Aliases
CG4871, D.SiaT, D.SialT, DSiaT, Dm ST, DmPST, DmSialT, Dmel\CG4871, Dmel_CG4871, ST, ST6Gal, SialT, St6Gal, dST6Gal I, dSiaT
Chromosome Location
Chr.2R: 24691422 - 24694386 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0035050
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_079129.3 NP_523853.1 5 - 6 1937 66
NM_166684.2 NP_726474.1 6 - 7 1317 66
GenBank mRNA AF218237.1 - 4 - 5 1319 66
AF397532.1 - 5 - 6 1280 66

View Full Product Details