Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rad23
Entrez Gene ID
43785
Gene Name
CG1836-PA
Gene Aliases
CG1836, DHR23, Dmel\CG1836, Dmel_CG1836, RAD23, rad23
Chromosome Location
Chr.4: 308093 - 310822 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0026777
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001272123.1 NP_001259052.1 4 - 5 1272 62
NM_143661.3 NP_651918.2 4 - 5 1272 62
NM_166752.2 NP_726561.1 4 - 5 1290 62
GenBank mRNA AF132147.1 - 4 - 5 1242 62

View Full Product Details