Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Asator
Entrez Gene ID
43794
Gene Name
Asator-PD
Gene Aliases
CG11533, CK I, CKI-like, Dmel\CG11533, Dmel_CG11533
Chromosome Location
Chr.4: 469409 - 488994 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0039908
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001272126.2 NP_001259055.1 8 - 9 2758 116
NM_001272127.1 NP_001259056.1 8 - 9 3345 116
NM_001272128.1 NP_001259057.1 8 - 9 3345 116
NM_001272129.1 NP_001259058.1 8 - 9 2824 116
NM_001297785.1 NP_001284714.1 8 - 9 2668 116
NM_143667.3 NP_651924.2 8 - 9 2681 116
NM_166765.3 NP_726576.2 8 - 9 2681 116
NM_166766.3 NP_726577.2 8 - 9 2824 116
GenBank mRNA BT133187.1 - 8 - 9 2647 116

View Full Product Details