Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gat
Entrez Gene ID
43805
Gene Name
GABA transporter
Gene Aliases
CG1732, CG1732a, DmGAT, Dmel\CG1732, Dmel_CG1732, GAT, anon-WO0149848.11, dGAT, gat
Chromosome Location
Chr.4: 629691 - 635350 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0039915
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_143673.3 NP_651930.2 4 - 5 839 109
NM_166779.4 NP_726594.2 4 - 5 837 109
GenBank mRNA BT010000.1 - 4 - 5 837 109

View Full Product Details