Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gw
Entrez Gene ID
43808
Gene Name
gawky
Gene Aliases
CG11484, CG31992, CG9905, Dmel\CG31992, Dmel_CG31992, GQ182, GW, GW182, Gw, Gw182, anon-EST:Posey205, dGW182, gw182
Chromosome Location
Chr.4: 649041 - 660608 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0051992
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014691.3 NP_001014691.2 4 - 5 2633 87
NM_166780.2 NP_726596.1 4 - 5 2877 87
NM_166781.2 NP_726597.1 4 - 5 2633 87
NM_166784.2 NP_726600.1 4 - 5 2516 87
NM_166785.2 NP_726601.1 4 - 5 2559 87
GenBank mRNA AY075429.1 - 4 - 5 2831 87

View Full Product Details