Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DAAM
Entrez Gene ID
31075
Gene Name
Dishevelled Associated Activator of Morphogenesis
Gene Aliases
CG14622, DAAM1, Daam, Dmel\CG14622, Dmel_CG14622, EG:114D9.2, anon-EST:Posey148, dDAAM, dDaam, daam
Chromosome Location
Chr.X: 1289713 - 1322362 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0025641
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001201593.2 NP_001188522.1 3 - 4 2294 62
NM_001272197.1 NP_001259126.1 4 - 5 2234 62
NM_001297842.1 NP_001284771.1 4 - 5 2147 62
NM_130544.4 NP_569900.3 2 - 3 1094 62
NM_166875.3 NP_726723.2 3 - 4 1977 62
NM_166876.3 NP_726724.1 5 - 6 2189 62

View Full Product Details