Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nmdar2
Entrez Gene ID
31107
Gene Name
NMDA receptor 2
Gene Aliases
79G8T, CG14793, CG14794, CG33513, CT34603, DmelNmdar2, Dmel\CG33513, Dmel_CG33513, DrNR2, EG:80H7.7, ESTS:79G8T, NMDA-R2, NMDAR, NMDAR2, NR2, anon-sts34, dNR2, dnmdar-II, dsNR2, nr2
Chromosome Location
Chr.X: 1483625 - 1507809 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0053513
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014714.2 NP_001014714.1 3 - 4 1853 86
NM_001014715.2 NP_001014715.1 2 - 3 1147 86
NM_001014716.2 NP_001014716.1 2 - 3 804 86
NM_001169164.1 NP_001162635.1 2 - 3 861 86
NM_001169165.1 NP_001162636.1 2 - 3 871 86
NM_001169166.1 NP_001162637.1 3 - 4 1854 86
NM_001297859.1 NP_001284788.1 3 - 4 1854 86
GenBank mRNA AY050490.1 - 2 - 3 804 86
AY050491.1 - 2 - 3 1147 86
AY616144.1 - 2 - 3 1128 86
AY616145.1 - 1 - 2 1386 86
AY616146.1 - 2 - 3 871 86
AY616147.1 - 2 - 3 861 86
AY616148.1 - 3 - 4 1854 86
AY616149.1 - 3 - 4 1854 86

View Full Product Details