Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
br
Entrez Gene ID
44505
Gene Name
broad
Gene Aliases
2B5, 2Bc, A18, BR, BR-C, BR-C Z1, BR-Z1, BR-Z2, BR-Z3, BR-Z4, BR-c, BRC, BRC-Z1, BRC-Z2, BRC-Z3, BRC-Z4, BR_C, Br, Br-C, Br-C Z2, Br-C-Z3, Br-Z2, Br-Z4, Br-c, BrC, BrC-Z1, BrZ3, CG11491, CG11509, CG11511, CG11514, Dmel\CG11491, Dmel_CG11491, EG:123F11.1, EG:17A9.1, EG:25D2.1, PP1, PP2, Z1, Z2, Z3, Z4, br-C, br-Z1, br-Z2, br-Z3, br-Z4, br-c, de12, ecs, l(1)2Ba, l(1)2Bab, l(1)2Bad, l(1)2Bb, l(1)2Bc, l(1)2Bd, l(1)ESHS5, l(1)G0018, l(1)G0042, l(1)G0284, l(1)G0284a, l(1)G0318, l(1)G0401, l(1)PP1, l(1)d norm-12, l(1)d.norm.1, l(1)dn1, l(1)n34, l(1)npr-1, l(1)npr1, l(1)pp-1, l(1)pp-2, l(1)pp1, l(1)pp2, l(1)ts132, l(1)ts144, l(1)ts358, l(1)ts376, npr, npr-1, npr1, nprl, nrp, nrp1, o.c.c., rbp, rdp, rds, uq
Chromosome Location
Chr.X: 1575132 - 1664869 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0283451
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144672.3 NP_001138144.1 2 - 3 1186 66
NM_001169167.2 NP_001162638.2 2 - 3 1069 66
NM_001201596.3 NP_001188525.1 2 - 3 1036 66
NM_001201597.1 NP_001188526.1 2 - 3 830 66
NM_001201598.1 NP_001188527.1 2 - 3 1163 66
NM_001258533.1 NP_001245462.1 3 - 4 1168 66
NM_001272205.1 NP_001259134.1 3 - 4 1116 66
NM_001382013.1 NP_001368968.1 2 - 3 1036 66
NM_080020.4 NP_524759.2 2 - 3 1036 66
NM_166893.3 NP_726749.2 2 - 3 1036 66
NM_166894.4 NP_726750.1 2 - 3 1036 66
NM_166895.2 NP_726751.2 2 - 3 862 66
NM_166896.3 NP_726752.1 2 - 3 1036 66
NM_166897.2 NP_726753.1 2 - 3 748 66
NM_166898.2 NP_726754.1 2 - 3 1036 66
GenBank mRNA AY069756.1 - 2 - 3 1036 66
BT125021.1 - 3 - 4 1168 66
X54665.1 - - 575 66

View Full Product Details