The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080301.3 | NP_525040.2 | 5 - 6 | 7138 | 61 |
NM_166911.4 | NP_726773.2 | 4 - 5 | 7201 | 61 |