Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
csw
Entrez Gene ID
45278
Gene Name
corkscrew
Gene Aliases
19-106, CG3954, CSW, Csw, Csw/Shp2, Dmel\CG3954, Dmel_CG3954, E(sev)1A, EG:BACN25G24.2, SHP-2, Shp-2, Shp2/csw, anon-WO03040301.207, anon-WO03040301.209, anon-WO03040301.219, csw/SHP-2, l(1)2Db, l(1)2Dd, l(1)G0170, l(1)GA114, l(1)csw
Chromosome Location
Chr.X: 2094234 - 2115701 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0000382
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001272245.1 NP_001259174.1 6 - 7 1485 62
NM_057782.5 NP_477130.1 6 - 7 963 62
NM_057783.3 NP_477131.1 4 - 5 1950 62
NM_166928.2 NP_726793.1 4 - 5 509 62
GenBank mRNA BT001484.1 - 6 - 7 897 62
BT149891.1 - 4 - 5 1754 62
M94730.1 - 6 - 7 932 62

View Full Product Details