Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
yin
Entrez Gene ID
31340
Gene Name
CG44402-PA
Gene Aliases
CG2913, CG34336, CG44402, Dmel\CG44402, Dmel_CG44402, EG:EG0002.1, OPT1, OpT1, Opt1, Yin, opt, opt1
Chromosome Location
Chr.X: 3862966 - 3871939 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0265575
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001297915.1 NP_001284844.1 5 - 6 622 75
NM_057799.4 NP_477147.2 3 - 4 704 75
NM_057800.4 NP_477148.2 3 - 4 526 75
NM_166993.2 NP_726894.1 3 - 4 517 75
GenBank mRNA BT031266.1 - 3 - 4 680 75

View Full Product Details