Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mdr49
Entrez Gene ID
36428
Gene Name
Multi drug resistance 49
Gene Aliases
CG3879, Dm5, Dmel\CG3879, Dmel_CG3879, MDR49, MDR5, Mes5, mdr49, mrd49
Chromosome Location
Chr.2R: 12940323 - 12946347 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0004512
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169661.1 NP_001163132.1 4 - 5 2775 68
NM_079000.4 NP_523724.2 7 - 8 3391 68
GenBank mRNA BT044206.1 - 7 - 8 3369 68
BT081353.1 - 4 - 5 2775 68

View Full Product Details